D-(-)-Diethyl tartrate
Leverancier: Molekula
MOLEM81782201EA
0
EUR
MOLEM81782201
D-(-)-Diethyl tartrate
D-(-)-Diethyl tartrate
Formule:
C₈H₁₄O₆ MW: 206,2 g/mol Kookpunt: 280 °C (1013 hPa) Smeltpunt: 17 °C Dichtheid: 1,2 g/cm³ (20 °C) |
MDL-nummer:
MFCD00064451 CAS-nummer: 13811-71-7 EINECS: 237-458-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...