(-)-Dimethyl-D-tartrate

Leverancier: Molekula

MOLE28005352-5GEA 0 EUR
MOLE28005352-5G
(-)-Dimethyl-D-tartrate
(-)-Dimethyl-D-tartrate
Formule: C₆H₁₀O₆
MW: 178,14 g/mol
Kookpunt: 245 °C (1013 hPa)
Smeltpunt: 45…48 °C
MDL-nummer: MFCD00008445
CAS-nummer: 13171-64-7
EINECS: 236-118-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR