1-Phenyl-2-tetrazoline-5-thione
Leverancier: Molekula
MOLE35197222-100GEA
0
EUR
MOLE35197222-100G
1-Phenyl-2-tetrazoline-5-thione
1-Phenyl-2-tetrazoline-5-thione
Formule:
C₇H₆N₄S MW: 178,22 g/mol Smeltpunt: 146…150 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00003129 CAS-nummer: 86-93-1 EINECS: 201-710-5 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...