Ethyl stearate for synthesis, Sigma-Aldrich®

Leverancier: Merck


M-Clarity™ quality level = MQ200

8.14352.5000EA 0 EUR
8.14352.5000 8.14352.9999
Ethyl stearate for synthesis, Sigma-Aldrich®
Ethyl stearate
Formule: C₂₀H₄₀O₂
MW: 312,54 g/mol
Kookpunt: 365 °C (1013 hPa)
Smeltpunt: 31 °C
Dichtheid: 0,8973 g/cm³ (25 °C)
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00009006
CAS-nummer: 111-61-5
EINECS: 203-887-4

Bestel nu

Specificaties testresultaten

Solubility (20 °C) insoluble
Melting Point 22 - 26 °C
Molar Mass 312.53 g/mol
Boiling Point 224 °C
Flash Point 112 °C
Refractive Index 1.4292 (40 °C, 589 nm)
Synonyms Stearic acid ethyl ester
Hill Formula C20H40O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR