Diethyl succinate, Supelco®

Leverancier: Merck


M-Clarity™ quality level = MQ100

07429-5ML-FEA 63.3 EUR
07429-5ML-F
Diethyl succinate, Supelco®
Diethyl succinate
Formule: C₈H₁₄O₄
MW: 174,2 g/mol
Kookpunt: 217 °C (1013 hPa)
Smeltpunt: –19 °C
Dichtheid: 1,04 g/cm³ (20 °C)
Vlampunt: 80 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00009208
CAS-nummer: 123-25-1
EINECS: 204-612-0
Merck Index: 12,09038

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR