Dimethyl glutarate

Leverancier: EHRENSTORFER GMBH

EHERC14034900EA 61.4 EUR
EHERC14034900
Dimethyl glutarate
Dimethyl glutarate
Formule: C₇H₁₂O₄
MW: 160,17 g/mol
Kookpunt: 211 °C (1013 hPa)
Smeltpunt: –37 °C
Dichtheid: 1,087 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00008468
CAS-nummer: 1119-40-0
EINECS: 214-277-2
Merck Index: 13,04486

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR