Calcium propionate
Leverancier: EHRENSTORFER GMBH
EHERC16493100EA
59.6
EUR
EHERC16493100
Calcium propionate
Calcium propionate
Formule:
(CH₃CH₂COO)₂Ca MW: 186,22 g/mol Smeltpunt: > 300 °C |
MDL-nummer:
MFCD00036137 CAS-nummer: 4075-81-4 EINECS: 223-795-8 Merck Index: 13,01703 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...