Copper(I) acetate 95%

Waarschuwing

ACRO378810010EA 31.6 EUR
ACRO378810010 ACRO378810050
Copper(I) acetate 95%
Copper(I) acetate
Formule: C₂H₃CuO₂
MW: 122,59 g/mol
Smeltpunt: 250 °C
MDL-nummer: MFCD00058908
CAS-nummer: 598-54-9
EINECS: 209-938-7
Merck Index: 12,02728

Bestel nu

Specificaties testresultaten

Appearance Light green to brown Powder and/or chunks
Infrared spectrum Conforms
Titration Complexometric 48.7 to 54.9 % (Cu)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR