Koper(II)acetaat monohydraat 98.0-102.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
1766-07EA
1378.32
EUR
1766-07
Koper(II)acetaat monohydraat 98.0-102.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Koper(II)acetaat monohydraat
Formule:
(H₃CCOO)₂Cu·H₂O MW: 199,65 g/mol Kookpunt: 240 °C (1013 hPa) Smeltpunt: 115 °C Dichtheid: 1,9 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00149570 CAS-nummer: 6046-93-1 EINECS: 205-553-3 UN: 3077 ADR: 9,III Merck Index: 12,02690 |
Specificaties testresultaten
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay ((CH₃COO)₂CuH₂O) | 98.0 - 102.0 % |
Insoluble Matter | ≤ 0.01 % |
pH of 5% Solution at 25°C | 5.0 - 6.0 |
Chloride (Cl) | ≤ 0.003 % |
Sulfate (SO₄) | ≤ 0.01 % |
Calcium (Ca) | ≤ 0.005 % |
Iron (Fe) | ≤ 0.002 % |
Nickel (Ni) | ≤ 0.01 % |
Potassium (K) | ≤ 0.01 % |
Sodium (Na) | ≤ 0.05 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...