1,18-Octadecanedioic acid
EHERC15710110EA
0
EUR
EHERC15710110
1,18-Octadecanedioic acid
1,18-Octadecanedioic acid
Formule:
C₁₈H₃₄O₄ MW: 314,47 g/mol |
MDL-nummer:
MFCD00142369 CAS-nummer: 871-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...