Thioureum ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Leverancier: Merck
|
Waarschuwing
|
M-Clarity™ quality level = MQ200
1.07979.1000EA
168
EUR
1.07979.1000
1.07979.0250
Thioureum ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Thioureum
Formule:
NH₂CSNH₂ MW: 76,12 g/mol Smeltpunt: 174…177 °C Dichtheid: 1,435 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00008067 CAS-nummer: 62-56-6 EINECS: 200-543-5 UN: 3077 ADR: 9,III Merck Index: 12,09505 |
Specificaties testresultaten
Solubility | 137 g/l (20 °C) |
Melting Point | 169 - 173 °C |
Molar Mass | 76.12 g/mol |
Bulk Density | 640 kg/m3 |
Vapor Pressure | (20 °C) |
Density | 1.405 g/cm3 (20 °C) |
pH | 6 - 8 (50 g/l, H2O, 20 °C) |
Ignition Point | 440 °C Dust |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous hazardous material |