Perfluoroicosane ≥97%

Leverancier: Thermo Scientific
Waarschuwing

L16828.03EA 62.4 EUR
L16828.03
Perfluoroicosane ≥97%
Perfluoroicosane
Formule: C₂₀F₄₂
MW: 1038,15 g/mol
Kookpunt: 320 °C
Smeltpunt: 162…165 °C
MDL-nummer: MFCD00000438
CAS-nummer: 37589-57-4
EINECS: 253-557-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR