Fluoraceetamide
Leverancier: EHRENSTORFER GMBH
|
Gevaar
|
EHERC13760000EA
107
EUR
EHERC13760000
Fluoraceetamide
Fluoraceetamide
Formule:
C₂H₄FNO MW: 77,06 g/mol Kookpunt: 259 °C (1013 hPa) Smeltpunt: 119 °C |
CAS-nummer:
640-19-7 EINECS: 211-363-1 Merck Index: 12,04204 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...