Pentafluoropropionic anhydride 98%

Gevaar

A14472.06EA 49.5 EUR
A14472.06 A14472.22 A14472.14
Pentafluoropropionic anhydride 98%
Pentafluoropropionic anhydride
Formule: C₆F₁₀O₃
MW: 310,05 g/mol
Kookpunt: 73…74 °C
Dichtheid: 1,589
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00000429
CAS-nummer: 356-42-3
EINECS: 206-604-2
UN: 3265
ADR: 8,II

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR