Iodoacetamide ≥98%, Proteomics Grade
Leverancier: VWR Chemicals
|
Waarschuwing
|
M216-30GEA
238
EUR
M216-30G
Iodoacetamide ≥98%, Proteomics Grade
Iodoacetamide
Formule:
ICH₂CONH₂ MW: 184,96 g/mol Kookpunt: 266 °C (1013 hPa) Smeltpunt: 91…94 °C Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00008028 CAS-nummer: 144-48-9 EINECS: 205-630-1 UN: 2928 ADR: 6.1,II |
Specificaties testresultaten
Loss on drying |
|
Melting point | 90 − 93 °C |
Protease | none detected |
Purity |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...