2-Diethylaminoethanol for synthesis, Sigma-Aldrich®
|
Gevaar
|
M-Clarity™ quality level = MQ200
8.03273.9027EA
334
EUR
8.03273.9027
8.03273.0100
8.03273.1000
8.03273.9999
2-Diethylaminoethanol for synthesis, Sigma-Aldrich®
2-Diethylaminoethanol
Formule:
C₆H₁₅NO MW: 117,19 g/mol Kookpunt: 162 °C (1013 hPa) Smeltpunt: –70 °C Dichtheid: 0,8921 g/cm³ (20 °C) Vlampunt: 52 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00002850 CAS-nummer: 100-37-8 EINECS: 202-845-2 UN: 2686 ADR: 8 (3),II Merck Index: 13,03139 |
Specificaties testresultaten
Solubility | (20 °C) soluble |
Melting Point | -70 °C |
Molar Mass | 117.19 g/mol |
Boiling Point | 163 °C (1013 hPa) |
Vapor Pressure | 1.9 hPa (20 °C) |
Flash Point | 51 °C |
Refractive Index | 1.4414 (20 °C, 589 nm) |
Explosion Limit | 0.7 - 10.1 %(V) |
Density | 0.88 g/cm3 (25 °C) |
pH | 11.5 (100 g/l, H2O, 20 °C) |
Ignition Point | 270 °C |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |