6-Aminohexan-1-ol

Leverancier: Molekula
Waarschuwing

MOLEM73615851EA 0 EUR
MOLEM73615851
6-Aminohexan-1-ol
6-Aminohexan-1-ol
Formule: C₆H₁₅NO
MW: 117,19 g/mol
Kookpunt: 227 °C (1013 hPa)
Smeltpunt: 53…55 °C
MDL-nummer: MFCD00008241
CAS-nummer: 4048-33-3
EINECS: 223-748-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR