N-Ethylethylenediamine 98+%

Leverancier: Thermo Scientific
Gevaar

ACRO155550500EA 140 EUR
ACRO155550500 ACRO155550050
N-Ethylethylenediamine 98+%
N-Ethylethylenediamine
Formule: C₄H₁₂N₂
MW: 88,15 g/mol
Kookpunt: 128 °C (1013 hPa)
Dichtheid: 0,83 g/cm³
Vlampunt: 10 °C
MDL-nummer: MFCD00008166
CAS-nummer: 110-72-5
EINECS: 203-795-4
UN: 2733
ADR: 3,II

Bestel nu

Specificaties testresultaten

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
GC ≥98.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR