Ethyl indol-3-ylacetate

EHERC14291000EA 89.3 EUR
EHERC14291000
Ethyl indol-3-ylacetate
Ethyl indol-3-ylacetate
Formule: C₁₂H₁₃NO₂
MW: 203,24 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005635
CAS-nummer: 778-82-5
EINECS: 212-296-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR