Methyl indol-3-ylacetate, Supelco®
M-Clarity™ quality level = MQ100
I9770-1GEA
74.5
EUR
I9770-1G
Methyl indol-3-ylacetate, Supelco®
Methyl indol-3-ylacetate
Formule:
C₁₁H₁₁NO₂ MW: 189,21 g/mol Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00022749 CAS-nummer: 1912-33-0 EINECS: 217-622-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...