(Indol-3-yl)acetic acid

Leverancier: EHRENSTORFER GMBH

EHERC14290000EA 127 EUR
EHERC14290000
(Indol-3-yl)acetic acid
(Indol-3-yl)acetic acid
Formule: C₁₀H₉NO₂
MW: 175,19 g/mol
Kookpunt: 120…125 °C (0,05 mmHg)
Smeltpunt: 167…170 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00005636
CAS-nummer: 87-51-4
EINECS: 201-748-2
Merck Index: 13,04986

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR