(4-Methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®
|
Gevaar
|
M-Clarity™ quality level = MQ200
8.21240.0250EA
169
EUR
8.21240.9999
8.21240.0250
8.21240.0100
(4-Methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®
(4-Methoxyphenyl)acetic acid
Formule:
(C₆H₄(OCH₃))CH₂COOH MW: 166,18 g/mol Kookpunt: 306 °C (1013 hPa) Smeltpunt: 84…86 °C Vlampunt: 193 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00004345 CAS-nummer: 104-01-8 EINECS: 203-166-4 |
Specificaties testresultaten
Solubility | 6 g/l (20 °C) |
Melting Point | 86 - 88 °C |
Molar Mass | 166.18 g/mol |
Bulk Density | 440 kg/m3 |
Boiling Point | 138 - 140 °C (4 hPa) |
Flash Point | 193 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...