Pempidine 95%
Leverancier: Thermo Scientific
|
Gevaar
|
ACRO416960050EA
81
EUR
ACRO416960050
Pempidine 95%
Pempidine
Formule:
C₁₀H₂₁N MW: 155,28 g/mol Kookpunt: 187…188 °C Smeltpunt: 11 °C Dichtheid: 0,858 g/cm³ Vlampunt: 50 °C |
MDL-nummer:
MFCD00006493 CAS-nummer: 79-55-0 EINECS: 201-211-2 UN: 1992 ADR: 3,III Merck Index: 12,07207 |
Specificaties testresultaten
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4570 to 1.4600 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...