Benzimidazole
Leverancier: EHRENSTORFER GMBH
EHERC10536500EA
83.7
EUR
EHERC10536500
Benzimidazole
Benzimidazole
Formule:
C₇H₆N₂ MW: 118,14 g/mol Kookpunt: 360 °C (1013 hPa) Smeltpunt: 170…173 °C Dichtheid: 1,23 g/cm³ (20 °C) |
MDL-nummer:
MFCD00005585 CAS-nummer: 51-17-2 EINECS: 200-081-4 Merck Index: 13,01082 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...