Dichlon, Supelco®

Leverancier: Merck
Waarschuwing


M-Clarity™ quality level = MQ100

35990-100G-FEA 37.7 EUR
35990-100G-F
Dichlon, Supelco®
Dichlon
Formule: C₁₀H₄Cl₂O₂
MW: 227,05 g/mol
Kookpunt: 304 °C (1013 hPa)
Smeltpunt: 193 °C
Dichtheid: 1,44 g/cm³ (16 °C)
MDL-nummer: MFCD00001677
CAS-nummer: 117-80-6
EINECS: 204-210-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,03071

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR