4-Cyanophenylboronic acid

Leverancier: Molekula
Waarschuwing

MOLE15798945-1GEA 0 EUR
MOLE15798945-1G
4-Cyanophenylboronic acid
4-Cyanophenylboronic acid
Formule: C₇H₆BNO₂
MW: 146,94 g/mol
Smeltpunt: > 300 °C
MDL-nummer: MFCD01318968
CAS-nummer: 126747-14-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR