5-Bromo-2-fluorobenzoic acid

Leverancier: Apollo Scientific
Waarschuwing

APOSPC4107-5GEA 12.6 EUR
APOSPC4107-5G
5-Bromo-2-fluorobenzoic acid
5-Bromo-2-fluorobenzoic acid
Formule: C₇H₄BrFO₂
MW: 219,01 g/mol
MDL-nummer: MFCD00143423
CAS-nummer: 146328-85-0
UN: 2811
ADR: 6.1,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR