O-Acetylsalicylic acid ≥99%, GPR RECTAPUR®
|
Waarschuwing
|
20110.290EA
116
EUR
20110.290
O-Acetylsalicylic acid ≥99%, GPR RECTAPUR®
O-Acetylsalicylic acid
Formule:
2-(CH₃CO₂)C₆H₄CO₂H MW: 180,16 g/mol Kookpunt: 140 °C (1013 hPa) Smeltpunt: 138…140 °C Dichtheid: 1,39 g/cm³ (20 °C) Vlampunt: 250 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00002430 CAS-nummer: 50-78-2 EINECS: 200-064-1 Merck Index: 13,00856 |
Specificaties testresultaten
Assay | Min. 99 % |
Heavy metals (as Pb) | Max. 10 ppm |
Ignition residue (SO4) | Max. 0.05 % |
Salicylic acid | Max. 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...