Bis(3,5,5-trimethylhexyl) phthalate for synthesis, Sigma-Aldrich®

Leverancier: Merck


M-Clarity™ quality level = MQ200

8.18375.0100EA 27.9 EUR
8.18375.0100 8.18375.1000
Bis(3,5,5-trimethylhexyl) phthalate for synthesis, Sigma-Aldrich®
Bis(3,5,5-trimethylhexyl) phthalate
Formule: C₂₆H₄₂O₄
MW: 418,62 g/mol
Dichtheid: 0,98 g/cm³ (20 °C)
CAS-nummer: 14103-61-8
EINECS: 237-954-4

Bestel nu

Specificaties testresultaten

Solubility (20 °C) insoluble
Melting Point -49 °C
Molar Mass 418.61 g/mol
Boiling Point 235 - 238 °C
Vapor Pressure  - 0.001 hPa (20 °C)
Flash Point 226 °C
Refractive Index 1.4819 (25 °C, 589 nm)
Explosion Limit 0.4 - 2.9 %(V)
Density 0.98 g/cm3 (20 °C)
pH 7 (H2O, 20 °C)
Ignition Point 305 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR