Dimethyl phthalate, TECHNICAL

25611.294EA 0 EUR
25611.294
Dimethyl phthalate, TECHNICAL
Dimethyl phthalate
Formule: C₁₀H₁₀O₄
MW: 194,19 g/mol
Kookpunt: 283,7 °C (1013 hPa)
Smeltpunt: 6 °C
Dichtheid: 1,192 g/cm³ (20 °C)
Vlampunt: 146 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00008425
CAS-nummer: 131-11-3
EINECS: 205-011-6
Merck Index: 13,03281

Bestel nu

Specificaties testresultaten

Identification Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR