N-phenylanthranilic acid for synthesis, Sigma-Aldrich®
Leverancier: Merck
M-Clarity™ quality level = MQ200
8.20979.0100EA 62.7 EUR
8.20979.0100 8.20979.9999
N-phenylanthranilic acid for synthesis, Sigma-Aldrich®
N-phenylanthranilic acid
Formule: C₁₃H₁₁NO₂
MW: 213,24 g/mol
Kookpunt: 375 °C (1013 hPa)
Smeltpunt: 182…185 °C
MDL-nummer: MFCD00002421
CAS-nummer: 91-40-7
EINECS: 202-066-8
Merck Index: 12,07427
Bestel nu
Specificaties testresultaten
Solubility (20 °C) insoluble
Melting Point 182 - 185 °C
Molar Mass 213.23 g/mol
Bulk Density 200 kg/m3