Diphenyl phthalate
Leverancier: LGC Standards PROMOCHEM
|
Waarschuwing
|
PROOCHE147EA
290
EUR
PROOCHE147
Diphenyl phthalate
Diphenyl phthalate
Formule:
C₂₀H₁₄O₄ MW: 318,33 g/mol Kookpunt: 405 °C (1013 hPa) Smeltpunt: 74…76 °C Dichtheid: 1,313 g/cm³ (20 °C) |
MDL-nummer:
MFCD00003038 CAS-nummer: 84-62-8 EINECS: 201-546-4 Merck Index: 12,07460 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...