2,4-Difluorobenzamide

Leverancier: Apollo Scientific

APOSPC2616-10GEA 88.1 EUR
APOSPC2616-25G APOSPC2616-10G APOSPC2616-50G
2,4-Difluorobenzamide
2,4-Difluorobenzamide
Formule: C₇H₅F₂NO
MW: 157,12 g/mol
MDL-nummer: MFCD00015547
CAS-nummer: 85118-02-1
EINECS: 285-654-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR