Pentafluorothiophenol 97%
|
Gevaar
|
ACRO129920010EA
34.9
EUR
ACRO129920010
ACRO129920050
ACRO129920250
Pentafluorothiophenol 97%
Pentafluorothiophenol
Formule:
C₆HF₅S MW: 200,13 g/mol Kookpunt: 143 °C (760 mmHg) Smeltpunt: –24 °C Dichtheid: 1,5 g/cm³ Vlampunt: 51 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00004828 CAS-nummer: 771-62-0 EINECS: 212-236-3 UN: 2920 ADR: 8,II |
Specificaties testresultaten
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4630 to 1.4650 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...