3-Aminothiophenol
Leverancier: Molekula
MOLEM18270207EA
0
EUR
MOLEM18270207
3-Aminothiophenol
3-Aminothiophenol
Formule:
C₆H₇NS MW: 125,19 g/mol Kookpunt: 110…112 °C (1 mmHg) Dichtheid: 1,1859 g/cm³ (20 °C) |
MDL-nummer:
MFCD00007791 CAS-nummer: 22948-02-3 EINECS: 245-344-4 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...