4-Allylanisole

Leverancier: EHRENSTORFER GMBH
Waarschuwing

EHERC10131000EA 74.4 EUR
EHERC10131000
4-Allylanisole
4-Allylanisole
Formule: C₁₀H₁₂O
MW: 148,2 g/mol
Kookpunt: 216 °C (1013 hPa)
Smeltpunt: 22,5 °C
Dichtheid: 0,965 g/cm³ (20 °C)
MDL-nummer: MFCD00008653
CAS-nummer: 140-67-0
EINECS: 205-427-8
Merck Index: 13,03740

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR