3-Bromo-2,6-diluorotoluene ≥99%

Leverancier: Thermo Scientific
Waarschuwing

H26159.MDEA 44.3 EUR
H26159.MD H26159.03
3-Bromo-2,6-diluorotoluene ≥99%
3-Bromo-2,6-diluorotoluene
Formule: C₇H₅BrF₂
MW: 207,02 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03412221
CAS-nummer: 221220-97-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR