6-Chloro-m-cresol 99%
Leverancier: Thermo Scientific
|
Gevaar
|
ACRO170415000EA
629
EUR
ACRO170415000
ACRO170410250
ACRO170411000
6-Chloro-m-cresol 99%
6-Chloro-m-cresol
Formule:
C₇H₇ClO MW: 142,58 g/mol Kookpunt: 196 °C (760 mmHg) Smeltpunt: 45…48 °C Vlampunt: 81 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00002175 CAS-nummer: 615-74-7 EINECS: 210-444-9 UN: 3437 ADR: 6.1,II |
Specificaties testresultaten
Appearance | White to yellow to beige Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 45 to 48 °C |
GC | ≥98.5 % |
HPLC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...