3'-Fluoroacetophenone
Leverancier: Molekula
|
Waarschuwing
|
MOLE61250889-5GEA
0
EUR
MOLE61250889-5G
3'-Fluoroacetophenone
3'-Fluoroacetophenone
Formule:
C₈H₇FO MW: 138,14 g/mol Kookpunt: 204 °C (1013 hPa) Smeltpunt: –3 °C Dichtheid: 1,12 g/cm³ (20 °C) |
MDL-nummer:
MFCD00000338 CAS-nummer: 455-36-7 EINECS: 207-245-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...