2-Fluoro-4-(trifluoromethyl)aniline ≥97%

Leverancier: Thermo Scientific
Gevaar

H26836.03EA 106 EUR
H26836.03 H26836.06
2-Fluoro-4-(trifluoromethyl)aniline ≥97%
2-Fluoro-4-(trifluoromethyl)aniline
Formule: C₇H₅F₄N
MW: 179,12 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01631629
CAS-nummer: 69409-98-9
UN: 2810
ADR: 6.1,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR