2-Fluoro-p-toluidine

Leverancier: Molekula

MOLE20149083-5GEA 0 EUR
MOLE20149083-5G
2-Fluoro-p-toluidine
2-Fluoro-p-toluidine
Formule: C₇H₈FN
MW: 125,15 g/mol
Kookpunt: 190 °C (1013 hPa)
Smeltpunt: 3 °C
Dichtheid: 1,1084 g/cm³ (18 °C)
MDL-nummer: MFCD00040975
CAS-nummer: 452-80-2
UN: 2810
ADR: 6.1,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR