4-Fluoro-m-toluidine
Leverancier: Molekula
|
Waarschuwing
|
MOLEM64185919EA
0
EUR
MOLEM64185919
4-Fluoro-m-toluidine
4-Fluoro-m-toluidine
Formule:
C₇H₈FN MW: 125,15 g/mol Kookpunt: 85…86 °C (9 mmHg) Smeltpunt: 36 °C Dichtheid: 1,12 g/cm³ (20 °C) |
MDL-nummer:
MFCD00025294 CAS-nummer: 452-69-7 EINECS: 207-207-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...