α,α,α-Trifluoro-p-cresol
Leverancier: Molekula
|
Waarschuwing
|
MOLEM80545511EA
0
EUR
MOLEM80545511
α,α,α-Trifluoro-p-cresol
α,α,α-Trifluoro-p-cresol
Formule:
C₇H₅F₃O MW: 162,11 g/mol Kookpunt: 185 °C (1013 hPa) Smeltpunt: 45…47 °C Dichtheid: 1,3 g/cm³ (25 °C) Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00002363 CAS-nummer: 402-45-9 EINECS: 206-945-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...