o-Tolyl isothiocyanate 99%
Leverancier: Thermo Scientific
|
Gevaar
|
ACRO292650250EA
143
EUR
ACRO292650250
ACRO292650050
o-Tolyl isothiocyanate 99%
o-Tolyl isothiocyanate
Formule:
C₈H₇NS MW: 149,22 g/mol Kookpunt: 239 °C (760 mmHg) Dichtheid: 1,11 g/cm³ Vlampunt: 107 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00004802 CAS-nummer: 614-69-7 EINECS: 210-390-6 UN: 2810 ADR: 6.1,II |
Specificaties testresultaten
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6351 to 1.6371 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...