3-Nitro-o-cresol

Leverancier: Molekula

MOLEM54449852EA 0 EUR
MOLEM54449852
3-Nitro-o-cresol
3-Nitro-o-cresol
Formule: C₇H₇NO₃
MW: 153,14 g/mol
Smeltpunt: 149…152 °C
MDL-nummer: MFCD00007241
CAS-nummer: 5460-31-1
EINECS: 226-739-0
UN: 2446
ADR: 6.1,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR