5-Methoxy-m-cresol

Leverancier: Molekula
Waarschuwing

MOLEM87126158EA 0 EUR
MOLEM87126158
5-Methoxy-m-cresol
5-Methoxy-m-cresol
Formule: C₈H₁₀O₂
MW: 138,17 g/mol
MDL-nummer: MFCD00059261
CAS-nummer: 3209-13-0
EINECS: 221-716-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR