sec-Pentylbenzene
EHERC15983200EA
0
EUR
EHERC15983200
sec-Pentylbenzene
sec-Pentylbenzene
Formule:
C₆H₅CH(CH₃)(CH₂)₂CH₃ MW: 148,25 g/mol Kookpunt: 199 °C (1013 hPa) Smeltpunt: –48 °C |
MDL-nummer:
MFCD00027145 CAS-nummer: 2719-52-0 EINECS: 220-324-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...