Phenol red, powder ACS pH-indicator
Leverancier: VWR Chemicals
0882-25GEA 115 EUR
0882-25G
Phenol red, powder ACS pH-indicator
Phenol red
Bright red powder
Formule: C₁₉H₁₄O₅S
MW: 354,38 g/mol
Smeltpunt: > 300 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00003552
CAS-nummer: 143-74-8
EINECS: 205-609-7
Merck Index: 12,07397
Bestel nu
Specificaties testresultaten
Solubility (0.1%, Alcohol) Pass
Visual Transition Interval (pH 6.8 - 8.2) Pass