Thymol blue

720-1050EA 82.8 EUR
720-1050
Thymol blue
Thymol blue
Formule: C₂₇H₃₀O₅S
MW: 466,6 g/mol
Smeltpunt: 221 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005869
CAS-nummer: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR