2,4-Dichloro-3,5-xylenol
Leverancier: EHRENSTORFER GMBH
EHERC12421500EA
116
EUR
EHERC12421500
2,4-Dichloro-3,5-xylenol
2,4-Dichloro-3,5-xylenol
Formule:
C₈H₈Cl₂O MW: 191,06 g/mol Kookpunt: 269 °C (1013 hPa) Smeltpunt: 96 °C |
MDL-nummer:
MFCD00019981 CAS-nummer: 133-53-9 EINECS: 205-109-9 Merck Index: 13,03102 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...